Tisk…
Ethylaluminium dichloride 25% in toluene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
192759-100G 192759-800G
SIAL192759-100GEA 1930 CZK
SIAL192759-100G SIAL192759-800G
Ethylaluminium dichloride 25% in toluene, Sigma-Aldrich®
Ethylaluminium dichloride
Vzorec: C₂H₅AlCl₂
MW: 126.95
MDL Number: MFCD00000457
CAS číslo: 563-43-9
EINECS: 209-248-6
UN: 3399
ADR: 4.3 I