Tisk…
Chromium diboride ≥99% (metals basis)
Dodavatel: Thermo Fisher Scientific
Synonyms: Chromium boride
12570.14 12570.22
12570.14EA 2460 CZK
12570.14 12570.22
Chromium diboride ≥99% (metals basis)
Chromium diboride
Purity: 99% CAS No.: 12007-16-8 Molecular Formula: CrB2 Formula Weight: 73.62 Physical Form: -325 Mesh Powder Odour: Odourless MDL No.: MFCD00016043
Vzorec: B₂Cr
MW: 73.62
Storage Temperature: Ambient
MDL Number: MFCD00016043
CAS číslo: 12007-16-8
EINECS: 234-499-3