Tisk…

Titanium diboride ≥99.5% (metals basis)

Dodavatel: Thermo Scientific

Synonyms: Titanium boride

44558.09 44558.18
44558.09EA 1390 CZK
44558.09 44558.18
Titanium diboride ≥99.5% (metals basis)
Titanium diboride

Purity: 99.5% CAS No.: 12045-63-5 Molecular Formula: B2Ti Formula Weight: 69.50 Physical Form: -325 Mesh Powder Density: 4.38 Appearance: Gray Odour: Odourless Melting Point: 3225° MDL No.: MFCD00049595

Vzorec: B₂Ti
MW: 69,5 g/mol
Bod tání: 3225 °C
Hustota: 4,38
Storage Temperature: Ambient
MDL Number: MFCD00049595
CAS číslo: 12045-63-5
EINECS: 234-961-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR