Tisk…

Chloromethyl methyl sulphide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

C54007-100G C54007-25G C54007-5G
SIALC54007-100GEA 13080 CZK
SIALC54007-100G SIALC54007-25G SIALC54007-5G
Chloromethyl methyl sulphide, Sigma-Aldrich®
Chloromethyl methyl sulphide
Vzorec: C₂H₅ClS
MW: 96,58 g/mol
MDL Number: MFCD00000923
CAS číslo: 2373-51-5
EINECS: 219-148-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR