Potassium heptafluorotantalate ≥99.99% (metals basis)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Potassium heptafluorotantalate(V)
43733.09
43733.30
43733.18
43733.09EA
3750
CZK
43733.09
43733.30
43733.18
Potassium heptafluorotantalate ≥99.99% (metals basis)
Potassium heptafluorotantalate
Purity: 99.99%
CAS No.: 16924-00-8
Molecular Formula: K2TaF7
Formula Weight: 392.14
Physical Form: Powder
Density: 4.560
UN No.: 3288
MDL No.: MFCD00042369
Vzorec:
F₇K₂Ta MW: 392,13 g/mol Hustota: 4,560 Storage Temperature: Ambient |
MDL Number:
MFCD00042369 CAS číslo: 16924-00-8 EINECS: 240-986-1 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...