Disodium terephthalate ≥99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Terephthalic acid disodium salt
Disodium terephthalate and its various derivatives are synthesized via simple acid-base chemistry for anode materials in Na ion batteries. They show excellent electrochemical performance, including little capacity fading over 90 cycles, ideal redox potential, and excellent rate performance, making them promising candidates for Na ion batteries.
Purity: 99+%
CAS No.: 10028-70-3
Molecular Formula: C6H4(COONa)2
Formula Weight: 210.10
Physical Form: Powder
Vzorec:
C₈H₄Na₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00013137 CAS číslo: 10028-70-3 EINECS: 233-075-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...