Tisk…

Dibenzyl azodicarboxylate 94%

Dodavatel: Thermo Scientific
Varování

Synonyms: Dibenzyl diazene-1,2-dicarboxylate

270270250 270270050
ACRO270270250EA 6510 CZK
ACRO270270250 ACRO270270050
Dibenzyl azodicarboxylate 94%
Dibenzyl azodicarboxylate
Vzorec: C₁₆H₁₄N₂O₄
MW: 298,3 g/mol
Bod tání: 44…52 °C
Bod vzplanutí: >230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016737
CAS číslo: 2449-05-0
EINECS: 219-508-0
UN: 1325
ADR: 4.1,III

Objednat

Výsledky specifikačního testu

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 44 to 52 °C
Loss on drying ≤0.75 %
UV spectrophotometry ≥92 %
Heavy metals (as Pb) ≤30 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR