Tisk…

Sodium hippurate 96%

Dodavatel: Thermo Fisher Scientific

A11723.18 A11723.30
A11723.18EA 2010 CZK
A11723.18 A11723.30
Sodium hippurate 96%
Sodium hippurate

Purity: 96% CAS No.: 532-94-5 Molecular Formula: C6H5CONHCH2CO2Na Formula Weight: 201.16 MDL No.: MFCD00002693

Vzorec: C₉H₈NNaO₃
Storage Temperature: Ambient
MDL Number: MFCD00002693
CAS číslo: 532-94-5
EINECS: 208-548-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR