N-(3-Pyridyl)thiourea ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-(3-Pyridyl)-2-thiourea, 1-(Pyridin-3-yl)thiourea
L01284.06
L01284.03
L01284.06EA
8030
CZK
L01284.06
L01284.03
N-(3-Pyridyl)thiourea ≥98%
N-(3-Pyridyl)thiourea
Purity: 98+%
CAS No.: 30162-37-9
Molecular Formula: C6H7N3S
Formula Weight: 153.21
Melting Point: 158-162°
Beilstein Reference: 472146
UN No.: 2811
MDL No.: MFCD00014635
Vzorec:
C₆H₇N₃S MW: 153,21 g/mol Bod tání: 158…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014635 CAS číslo: 30162-37-9 EINECS: 250-075-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...