Tisk…

(2-Naphthoxy)acetic acid ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Naphthoxyacetic acid

A15412.22 A15412.36
A15412.22EA 1270 CZK
A15412.22 A15412.36
(2-Naphthoxy)acetic acid ≥97%
(2-Naphthoxy)acetic acid

Purity: 97% CAS No.: 120-23-0 Molecular Formula: C12H10O3 Formula Weight: 202.21 Melting Point: 155-157° Beilstein Reference: 1074148 Merck Reference: 14,6397 MDL No.: MFCD00004066

Vzorec: C₁₂H₁₀O₃
MW: 202,21 g/mol
Bod tání: 155…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00004066
CAS číslo: 120-23-0
EINECS: 204-380-0
Merck Index: 14,6397

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR