Tisk…

Di-tert-butyl azodicarboxylate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Di-tert-butyl diazene-1,2-dicarboxylate

M-Clarity™ quality level = MQ200

11625-25G-F 11625-5G-F
SIAL11625-25G-FEA 6090 CZK
SIAL11625-25G-F SIAL11625-5G-F
Di-tert-butyl azodicarboxylate, Sigma-Aldrich®
Di-tert-butyl azodicarboxylate
Vzorec: C₁₀H₁₈N₂O₄
MW: 230,26 g/mol
MDL Number: MFCD00015001
CAS číslo: 870-50-8
EINECS: 212-796-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR