Di-tert-butyl azodicarboxylate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Di-tert-butyl diazene-1,2-dicarboxylate
L00294.06
L00294.14
L00294.06EA
2020
CZK
L00294.06
L00294.14
Di-tert-butyl azodicarboxylate ≥98%
Di-tert-butyl azodicarboxylate
Purity: 98%
CAS No.: 870-50-8
Molecular Formula: (CH3)3CO2C-N=N-CO2C(CH3)3
Formula Weight: 230.27
Melting Point: 89-92°
Beilstein Reference: 1911434
Vzorec:
C₁₀H₁₈N₂O₄ MW: 230.26 Bod tání: 89…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015001 CAS číslo: 870-50-8 EINECS: 212-796-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...