Tisk…
Sarcosine methyl ester hydrochloride ≥98%
Dodavatel: Thermo Scientific
Varování
Synonyms: H-Sar-OMe·HCl, N-Sar-OMe·HCl, Methyl N-methylglycinate hydrochloride
L08572.09 L08572.18
L08572.09EA 1180 CZK
L08572.09 L08572.18
Sarcosine methyl ester hydrochloride ≥98%
Sarcosine methyl ester hydrochloride
Purity: 98% CAS No.: 13515-93-0 Molecular Formula: CH3NHCH2CO2CH3.HCl Formula Weight: 139.58 Melting Point: ca 105° dec. Beilstein Reference: 3677128
Vzorec: C₄H₉NO₂·HCl
MW: 139,58 g/mol
Bod tání: ∼ 105 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00038876
CAS číslo: 13515-93-0
EINECS: 236-853-2