Sarcosine ethyl ester hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Sarcosine Ethyl Ester HCl, H-Sar-OEt·HCl, Ethyl 2-(methylamino)acetate hydrochloride
M-Clarity™ quality level = MQ200
255084-10G
255084-50G
SIAL255084-10GEA
1940
CZK
SIAL255084-10G
SIAL255084-50G
Sarcosine ethyl ester hydrochloride, Sigma-Aldrich®
Sarcosine ethyl ester hydrochloride
Vzorec:
C₅H₁₁NO₂·HCl MW: 153,61 g/mol |
MDL Number:
MFCD00012506 CAS číslo: 52605-49-9 EINECS: 258-037-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...