Tisk…

Potassium ethyl malonate ≥98%

Dodavatel: Thermo Scientific

A10720.14 A10720.22 A10720.36
A10720.14EA 1640 CZK
A10720.14 A10720.22 A10720.36
Potassium ethyl malonate ≥98%
Potassium ethyl malonate
Vzorec: C₅H₇KO₄
Bod tání: ∼ 200 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00035603
CAS číslo: 6148-64-7
EINECS: 228-156-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR