Tisk…

Nonadecanoic acid methyl ester, Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

74208-1G 74208-5G
74208-1GEA 4010 CZK
74208-1G 74208-5G
Nonadecanoic acid methyl ester, Supelco®
Nonadecanoic acid methyl ester
Vzorec: C₂₀H₄₀O₂
MDL Number: MFCD00009011
CAS číslo: 1731-94-8
EINECS: 217-056-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fd3fe6c8ec159c2920d00adf28974c5c62dd03f&vid=a031dd9640f2db91d675d7c4ffdc386e4c1246fd&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR