Nitrilotriacetic acid trisodium salt monohydrate, Sigma-Aldrich®
Dodavatel: Merck
72565-1KG
72565-250G
SIAL72565-1KGEA
3130
CZK
SIAL72565-1KG
SIAL72565-250G
Nitrilotriacetic acid trisodium salt monohydrate, Sigma-Aldrich®
Nitrilotriacetic acid trisodium salt monohydrate
Vzorec:
C₆H₆NNa₃O₆·1H₂O |
MDL Number:
MFCD00150789 CAS číslo: 18662-53-8 EINECS: 225-768-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...