Tisk…

Nitrilotriacetic acid trisodium salt monohydrate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Nitrilotriacetic acid, trisodium salt, monohydrate

A18348.22 A18348.36
A18348.22EA 996 CZK
A18348.22 A18348.36
Nitrilotriacetic acid trisodium salt monohydrate ≥98%
Nitrilotriacetic acid trisodium salt monohydrate

Purity: 98+% CAS No.: 18662-53-8 Molecular Formula: N(CH2CO2Na)3.H2O Formula Weight: 275.12 (257.10anhy) Melting Point: 300° Beilstein Reference: 3774022 Merck Reference: 14,6579 MDL No.: MFCD00150789

Vzorec: C₆H₆NNa₃O₆·1H₂O
MW: 275,1 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150789
CAS číslo: 18662-53-8
EINECS: 205-355-7
Merck Index: 14,6579

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR