Tisk…

N-Acetyl-L-glutamic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

855642-100G 855642-25G
SIAL855642-100GEA 3200 CZK
SIAL855642-100G SIAL855642-25G
N-Acetyl-L-glutamic acid, Sigma-Aldrich®
N-Acetyl-L-glutamic acid
Vzorec: C₇H₁₁NO₅
MW: 189.17
MDL Number: MFCD00002802
CAS číslo: 1188-37-0
EINECS: 214-708-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR