Diethyl azodicarboxylate 40% ∼2.2 mol/l in toluene, Sigma-Aldrich®
Dodavatel: Merck
11627-250ML-F
11627-50ML-F
SIAL11627-250ML-FEA
13340
CZK
SIAL11627-250ML-F
SIAL11627-50ML-F
Diethyl azodicarboxylate 40% ∼2.2 mol/l in toluene, Sigma-Aldrich®
Diethyl azodicarboxylate
Vzorec:
C₆H₁₀N₂O₄ MW: 174.16 |
MDL Number:
MFCD00009103 CAS číslo: 1972-28-7 EINECS: 217-821-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...