Triethylenetetramine mixture of isomers, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
132098-100G
132098-18KG
132098-1KG
132098-500G
132098-2KG
SIAL132098-100GEA
636
CZK
SIAL132098-100G
SIAL132098-18KG
SIAL132098-1KG
SIAL132098-500G
SIAL132098-2KG
Triethylenetetramine mixture of isomers, Sigma-Aldrich®
Triethylenetetramine
Vzorec:
C₆H₁₈N₄ |
MDL Number:
MFCD00008169 CAS číslo: 112-24-3 EINECS: 203-950-6 UN: 2259 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...