Tisk…

Arecolinium bromide

Dodavatel: Cayman Chemical

13662-100 13662-250 13662-500
CAYM13662-100EA 797 CZK
CAYM13662-100 CAYM13662-250 CAYM13662-500
Arecolinium bromide
Arecolinium bromide
Vzorec: C₈H₁₄BrNO₂
MDL Number: MFCD00039041
CAS číslo: 300-08-3
EINECS: 206-087-3
UN: 1544
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR