Tisk…

Diammonium wolframate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ammonium tungstate

M-Clarity™ quality level = MQ100

322385-10G 322385-50G
SIAL322385-10GEA 3910 CZK
SIAL322385-10G SIAL322385-50G
Diammonium wolframate, Sigma-Aldrich®
Diammonium wolframate
Vzorec: H₈N₂O₄W
MW: 283,91 g/mol
MDL Number: MFCD00015956
CAS číslo: 11140-77-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR