Diammonium wolframate ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
Synonyms:
Ammonium tungstate
44588.09
44588.18
44588.30
44588.09EA
3210
CZK
44588.09
44588.18
44588.30
Diammonium wolframate ≥99.99% (metals basis)
Diammonium wolframate
Purity: 99.99+%
CAS No.: 11140-77-5
Molecular Formula: (NH4)2WO4
Formula Weight: 283.91
Physical Form: Powder
Solubility: Soluble in warm water
MDL No.: MFCD00015956
Vzorec:
H₈N₂O₄W MW: 283.91 Storage Temperature: Ambient |
MDL Number:
MFCD00015956 CAS číslo: 11140-77-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...