Tisk…

trans,trans-2,4-Nonadienal, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W321206-100G-K W321206-1KG-K W321206-25G-K
SIALW321206-100G-KEA 8850 CZK
SIALW321206-100G-K SIALW321206-1KG-K SIALW321206-25G-K
trans,trans-2,4-Nonadienal, Sigma-Aldrich®
trans,trans-2,4-Nonadienal
Vzorec: C₉H₁₄O
MDL Number: MFCD00007006
CAS číslo: 5910-87-2
EINECS: 227-629-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR