trans,trans-2,4-Hexadienal, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
180343-100ML
180343-25ML
SIAL180343-100MLEA
5350
CZK
SIAL180343-100ML
SIAL180343-25ML
trans,trans-2,4-Hexadienal, Sigma-Aldrich®
trans,trans-2,4-Hexadienal
Vzorec:
C₆H₈O MW: 96.13 |
MDL Number:
MFCD00007004 CAS číslo: 142-83-6 EINECS: 205-564-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...