Tisk…

trans,trans-2,4-Heptadienal, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2E,4E)-Hepta-2,4-dienal

M-Clarity™ quality level = MQ300

W316407-100G-K W316407-1KG-K
SIALW316407-100G-KEA 8390 CZK
SIALW316407-100G-K SIALW316407-1KG-K
trans,trans-2,4-Heptadienal, Sigma-Aldrich®
trans,trans-2,4-Heptadienal
Vzorec: C₇H₁₀O
MDL Number: MFCD00007005
CAS číslo: 4313-03-5
EINECS: 224-328-0
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR