Tisk…
6,7-Dimethoxy-1-tetralone ≥97%
Dodavatel: Thermo Fisher Scientific
Synonyms: 6,7-Dimethoxy-3,4-dihydronaphthalen-1(2H)-one
B25226.03 B25226.06 B25226.14
B25226.03EA 1750 CZK
B25226.03 B25226.06 B25226.14
6,7-Dimethoxy-1-tetralone ≥97%
6,7-Dimethoxy-1-tetralone
Purity: 97% CAS No.: 13575-75-2 Molecular Formula: C12H14O3 Formula Weight: 206.24 Melting Point: 98-100° MDL No.: MFCD00134100
Vzorec: C₁₂H₁₄O₃
MW: 206.24
Bod tání: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00134100
CAS číslo: 13575-75-2