Tisk…

6-Methoxy-2-tetralone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Methoxy-3,4-dihydronaphthalen-2(1H)-one, 3,4-Dihydro-6-methoxynaphthalen-2(1H)-one

M-Clarity™ quality level = MQ100

184063-1G 184063-5G
SIAL184063-1GEA 4010 CZK
SIAL184063-1G SIAL184063-5G
6-Methoxy-2-tetralone, Sigma-Aldrich®
6-Methoxy-2-tetralone
Vzorec: C₁₁H₁₂O₂
MW: 176,22 g/mol
MDL Number: MFCD00001729
CAS číslo: 2472-22-2
EINECS: 219-592-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR