Tisk…

6-Amino-1-tetralone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Amino-3,4-dihydronaphthalen-1(2H)-one, 6-Amino-1,2,3,4-tetrahydronaphthalen-1-one

M-Clarity™ quality level = MQ100

678422-10G 678422-1G
SIAL678422-10GEA 8560 CZK
SIAL678422-10G SIAL678422-1G
6-Amino-1-tetralone, Sigma-Aldrich®
6-Amino-1-tetralone
Vzorec: C₁₀H₁₁NO
MW: 161,2 g/mol
MDL Number: MFCD00099462
CAS číslo: 3470-53-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR