Tisk…

1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,4-Dihydronaphthalen-1(2H)-one

M-Clarity™ quality level = MQ100

T19003-100G T19003-500G T19003-5G
SIALT19003-100GEA 2000 CZK
SIALT19003-100G SIALT19003-500G SIALT19003-5G
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone), Sigma-Aldrich®
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone)
Vzorec: C₁₀H₁₀O
MDL Number: MFCD00001688
CAS číslo: 529-34-0
EINECS: 208-460-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR