Tisk…

7-Bromo-1-tetralone ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: 7-Bromo-3,4-dihydronaphthalen-1(2H)-one

H50604.03 H50604.06
H50604.03EA 2850 CZK
H50604.03 H50604.06
7-Bromo-1-tetralone ≥97%
7-Bromo-1-tetralone

Purity: 97% CAS No.: 32281-97-3 Molecular Formula: C10H9BrO Formula Weight: 225.09 Melting Point: 73-77° MDL No.: MFCD02179287

Vzorec: C₁₀H₉BrO
MW: 225.08
Bod tání: 73…77 °C
Storage Temperature: Ambient
MDL Number: MFCD02179287
CAS číslo: 32281-97-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR