6-Bromo-2-tetralone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
6-Bromo-3,4-dihydronaphthalen-2(1H)-one
M-Clarity™ quality level = MQ100
639885-1G
639885-5G
SIAL639885-1GEA
3950
CZK
SIAL639885-1G
SIAL639885-5G
6-Bromo-2-tetralone, Sigma-Aldrich®
6-Bromo-2-tetralone
Vzorec:
C₁₀H₉BrO MW: 225,08 g/mol |
MDL Number:
MFCD00239388 CAS číslo: 4133-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...