2-Oxazolidinone ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Oxazolidone, Oxazolidin-2-one
A14543.14
A14543.22
A14543.36
AB118367.0025
AB118367.0100
A14543.14EA
1580
CZK
A14543.14
A14543.22
A14543.36
ABCRAB118367.0025
ABCRAB118367.0100
2-Oxazolidinone ≥99%
2-Oxazolidinone
Purity: 99%
CAS No.: 497-25-6
Molecular Formula: C3H5NO2
Formula Weight: 87.08
Melting Point: 84-89°
Boiling Point: 220°/48mm
MDL No.: MFCD00005268
Vzorec:
C₃H₅NO₂ Bod varu: 220 °C (48 mmHg) Bod tání: 84…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005268 CAS číslo: 497-25-6 EINECS: 207-840-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...