Tisk…

2(5H)-Furanone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Furan-2(5H)-one

M-Clarity™ quality level = MQ200

283754-1G 283754-5G
SIAL283754-1GEA 3010 CZK
SIAL283754-1G SIAL283754-5G
2(5H)-Furanone, Sigma-Aldrich®
2(5H)-Furanone
Vzorec: C₄H₄O₂
MW: 84,07 g/mol
MDL Number: MFCD00005376
CAS číslo: 497-23-4
EINECS: 207-839-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR