Tisk…

Potassium dicyanoaurate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Gold(I) potassium cyanide, Potassium dicyanoaurate(I)

M-Clarity™ quality level = MQ100

298115-1G 298115-5G
SIAL298115-1GEA 6120 CZK
SIAL298115-1G SIAL298115-5G
Potassium dicyanoaurate, Sigma-Aldrich®
Potassium dicyanoaurate
Vzorec: C₂AuKN₂
MDL Number: MFCD00011414
CAS číslo: 13967-50-5
EINECS: 237-748-4
UN: 1588
ADR: 6.1,I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR