Tisk…

Triethylborane 1 mol/l THF, Sigma-Aldrich®

Dodavatel: Merck


MQ100

179701-100ML 179701-800ML
SIAL179701-100MLEA 3150 CZK
SIAL179701-100ML SIAL179701-800ML
Triethylborane 1 mol/l THF, Sigma-Aldrich®
Triethylborane
Vzorec: C₆H₁₅B
MW: 98
MDL Number: MFCD00009022
CAS číslo: 97-94-9
EINECS: 202-620-9
UN: 2924
ADR: 3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR