Hexamethyldisilathiane, tech.
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Bis(trimethylsilyl)sulphide
44818.03
44818.06
44818.03EA
1550
CZK
44818.03
44818.06
Hexamethyldisilathiane, tech.
Hexamethyldisilathiane
CAS No.: 3385-94-2
Molecular Formula: C6H18SSi2
Formula Weight: 178.44
Flash Point: 26°(79°F)
Physical Form: Liquid in ampoules
Density: 0.851
Refractive Index: 1.4598
Sensitivity: Air Sensitive
Appearance: Colourless
Odour: Pungent
Boiling Point: 162-166°
Beilstein Reference: 1740046
UN No.: 1993
MDL No.: MFCD00014851
Vzorec:
C₆H₁₈SSi₂ MW: 178.45 Bod varu: 162…166 °C Hustota: 0.851 Bod vzplanutí: 26 °C (79 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014851 CAS číslo: 3385-94-2 EINECS: 222-201-4 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...