2-(2-Naphthoxy)ethanol 98+%
Dodavatel: Thermo Scientific
Synonyms:
2-(Naphthalen-2-yloxy)ethanol, 2-(2-Naphthyloxy)ethanol, Anavenol
A11636.06
A11636.14
A11636.22
A11636.06EA
2240
CZK
A11636.06
A11636.14
A11636.22
2-(2-Naphthoxy)ethanol 98+%
2-(2-Naphthoxy)ethanol
Purity: 98+%
CAS No.: 93-20-9
Molecular Formula: C12H12O2
Formula Weight: 188.23
Melting Point: 72-76°
Beilstein Reference: 2086973
Merck Reference: 14,6413
MDL No.: MFCD00016809
Vzorec:
C₁₂H₁₂O₂ MW: 188,23 g/mol Bod tání: 72…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016809 CAS číslo: 93-20-9 EINECS: 202-228-8 Merck Index: 14,6413 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...