Diethyl propylmalonate ≥99%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
Diethyl 2-propylmalonate
B23304.14
B23304.22
B23304.14EA
1470
CZK
B23304.14
B23304.22
Diethyl propylmalonate ≥99%
Diethyl propylmalonate
Purity: 99%
CAS No.: 2163-48-6
Molecular Formula: C10H18O4
Formula Weight: 202.25
Flash Point: 91°(197°F)
Refractive Index: 1.4205
Boiling Point: 221-222°
MDL No.: MFCD00009168
Vzorec:
C₁₀H₁₈O₄ MW: 202,25 g/mol Bod varu: 221…222 °C Hustota: 0,987 Bod vzplanutí: 91 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009168 CAS číslo: 2163-48-6 EINECS: 218-492-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...