Diethyl methylmalonate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
126136-100G
126136-500G
SIAL126136-100GEA
2220
CZK
SIAL126136-100G
SIAL126136-500G
Diethyl methylmalonate, Sigma-Aldrich®
Diethyl methylmalonate
Vzorec:
C₈H₁₄O₄ |
MDL Number:
MFCD00009162 CAS číslo: 609-08-5 EINECS: 210-175-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...