Diethyl methylmalonate ≥99%
Dodavatel: Thermo Fisher Scientific
Varování
|
Synonyms:
Diethyl 2-methylmalonate
A12078.0B
A12078.18
A12078.30
A12078.0BEA
9510
CZK
A12078.0B
A12078.18
A12078.30
Diethyl methylmalonate ≥99%
Diethyl methylmalonate
Purity: 99%
CAS No.: 609-08-5
Molecular Formula: C8H14O4
Formula Weight: 174.20
Flash Point: 85°(185°F)
Refractive Index: 1.4130
Boiling Point: 198-199°
Beilstein Reference: 637028
MDL No.: MFCD00009162
Vzorec:
C₈H₁₄O₄ Bod varu: 198…199 °C Hustota: 1,02 Bod vzplanutí: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009162 CAS číslo: 609-08-5 EINECS: 210-175-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...