Tisk…

Diethyl allylmalonate 96%

Dodavatel: Thermo Fisher Scientific
Varování

L02286.14 L02286.22
L02286.14EA 1340 CZK
L02286.14 L02286.22
Diethyl allylmalonate 96%
Diethyl allylmalonate

Purity: 97% CAS No.: 2049-80-1 Molecular Formula: C10H16O4 Formula Weight: 200.23 Flash Point: 92°(197°F) Refractive Index: 1.4310 Boiling Point: 222-223° Beilstein Reference: 1709466 MDL No.: MFCD00009155

Vzorec: C₁₀H₁₆O₄
Bod varu: 222…223 °C
Hustota: 1,016
Bod vzplanutí: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009155
CAS číslo: 2049-80-1
EINECS: 218-072-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR