Hafnium dichloride oxide octahydrate ≥98% (metals basis excluding Zr) (max. 1.5% Zr)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Hafnium(IV) dichloride oxide octahydrate
11833.14
11833.22
11833.36
11833.14EA
2760
CZK
11833.14
11833.22
11833.36
Hafnium dichloride oxide octahydrate ≥98% (metals basis excluding Zr) (max. 1.5% Zr)
Hafnium dichloride oxide octahydrate
Purity: 98+%
CAS No.: 14456-34-9
Molecular Formula: Cl2HfO.8H2O
Formula Weight: 409.52 (265.40anhy)
Physical Form: Powder
Density: 2.2
Appearance: White
Odour: Pungent
Solubility: Soluble in water
UN No.: 3260
MDL No.: MFCD00150417
Synthesis of Hafnium (IV) oxide
Vzorec:
Cl₂HfO·8H₂O MW: 409,52 g/mol Hustota: 2,2 Storage Temperature: Ambient |
MDL Number:
MFCD00150417 CAS číslo: 14456-34-9 EINECS: 237-345-3 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...