Tisk…
Cadmium perchlorate hexahydrate
Dodavatel: Thermo Fisher Scientific
Varování
12936.18 12936.30
12936.18EA 2890 CZK
12936.18 12936.30
Cadmium perchlorate hexahydrate
Cadmium perchlorate hexahydrate
CAS No.: 10326-28-0 Molecular Formula: Cd(ClO4)2.6H2O Formula Weight: 419.39 (311.30anhy) Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00015992
Vzorec: CdCl₂O₈·6H₂O
MW: 419.4
Storage Temperature: Ambient
MDL Number: MFCD00150202
CAS číslo: 10326-28-0
UN: 1481
ADR: 5.1 II