Phthaloyl dichloride ≥94%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L08753.22
L08753.36
L08753.22EA
1440
CZK
L08753.22
L08753.36
Phthaloyl dichloride ≥94%
Phthaloyl dichloride
Purity: 94%
CAS No.: 88-95-9
Molecular Formula: C8H4Cl2O2
Formula Weight: 203.03
Flash Point: 151°(304°F)
Refractive Index: 1.5680
Melting Point: 12°
Boiling Point: 276°
Beilstein Reference: 608200
Merck Reference: 14,7375
UN No.: 3265
MDL No.: MFCD00000666
Vzorec:
C₈H₄Cl₂O₂ MW: 203,02 g/mol Bod varu: 276 °C Bod tání: 12 °C Hustota: 1,409 Bod vzplanutí: 151 °C (304 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000666 CAS číslo: 88-95-9 EINECS: 201-869-0 UN: 3265 ADR: 8,II Merck Index: 14,7375 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...