Tisk…

Chloroform : isoamyl alcohol (24:1) (v/v), Supelco®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

25666-100ML 25666-500ML
SUPL25666-100MLEA 2740 CZK
SUPL25666-100ML SUPL25666-500ML
Chloroform : isoamyl alcohol (24:1) (v/v), Supelco®
Chloroform : isoamyl alcohol
Vzorec: CHCl₃,C₅H₁₂O
Storage Temperature: Ambient
UN: 1888
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR