Europium(III) sulphate octahydrate ≥99.9%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
29106.04
29106.09
29106.04EA
2540
CZK
29106.04
29106.09
Europium(III) sulphate octahydrate ≥99.9%
Europium(III) sulphate octahydrate
Purity: 99.9%
CAS No.: 10031-55-7
Molecular Formula: Eu2(SO4)3.8H2O
Formula Weight: 736.23 (592.10anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Merck Reference: 14,3904
MDL No.: MFCD00149704
Vzorec:
Eu₂O₁₂S₃·8H₂O MW: 736.23 Bod tání: 375 °C (-8H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00149703 CAS číslo: 10031-55-7 EINECS: 236-903-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...