Isopropylmalonic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L10171.06
L10171.14
L10171.06EA
2360
CZK
L10171.06
L10171.14
Isopropylmalonic acid ≥98%
Isopropylmalonic acid
Purity: 98+%
CAS No.: 601-79-6
Molecular Formula: C6H10O4
Formula Weight: 146.14
Melting Point: 88-90°
Beilstein Reference: 1723234
Vzorec:
C₆H₁₀O₄ Bod tání: 88…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021717 CAS číslo: 601-79-6 EINECS: 210-008-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...