2-Amino-4-chloropyrimidine 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H55007.03
H55007.06
H55007.03EA
2250
CZK
H55007.03
H55007.06
2-Amino-4-chloropyrimidine 98%
2-Amino-4-chloropyrimidine
Purity: 98%
CAS No.: 3993-78-0
Molecular Formula: C4H4ClN3
Formula Weight: 129.55
Storage Temperature: 2°C to 8°C
Melting Point: 155-160° dec.
MDL No.: MFCD00038021
Vzorec:
C₄H₄ClN₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00038021 CAS číslo: 3993-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...